Shipping bottles, stainless steel, UN approved, with screw cap, DURAN®
Leverandør: DWK Life Sciences
299016055
215-3263EA 12350 NOK
215-3263
Shipping bottles, stainless steel, UN approved, with screw cap, DURAN®
Flasker Metallflasker
Manufactured from corrosion resistant AISI Type 316 L (1.4404) stainless steel, and hygienically constructed with all welded seams.
  • Container for shipping hazardous or non hazardous liquids
  • Storage or shipping of precious liquids, such as high purity fine chemicals, pharmaceutical or cosmetic products
  • Storage of light-sensitive liquids
This unbreakable steel bottle is ideal for the storage and safe shipping of dangerous liquid goods such as solvents and reagents without the need for additional protective outer packaging. The bottle has a smooth innerMore Product Information