Laboratorieflasker, smal åpning, med gjenge, DURAN®
Leverandør: DWK Life Sciences
218105403 218165409 1092234 1092235 1094368 1094367 218102406 218162403
215-0042EA 595 NOK
215-0042 215-3268 215-3264 215-3265 215-3267 215-3266 215-3881 215-4190
Laboratorieflasker, smal åpning, med gjenge, DURAN®
Flasker Glassflasker
Bottles, pressure plus+, DURAN® borosilicate glass 3.3, clear. Safe working under pressure or vacuum, sampling under pressure, storage of gas generating media. With easy to read scale and large labelling field for easy marking, in fired-on, highly durable white ceramic.
  • Blue scale for visual differentiation from the standard laboratory bottle
  • Available in clear or amber glass
  • Glass Type I according to USP, EP and JP
  • Amber glass meets USP 671 and Eur. Ph. 3.2.1. for protection against UV light
Pressure resistance conforms to DIN ISO 1595. Vacuum and/orMore Product Information