Jars, with pressure seal cap
Leverandør: LAMAPLAST
54 28 29
215-4060EA 53 NOK
215-4060 215-4062 215-4063
Jars, with pressure seal cap
Krukker Bokser, generell bruk
Jars, made of HDPE, with pressure sealing cap.
  • Pressure cap is tamper proof
  • Suitable for food storage
  • Good chemical and temperature resistance
Ideal for the storage of creams, semi-solid substances, granulates and powders.