VWR®, Laboratory Bottles, Coated
Leverandør: VWR Collection
215-4267EA 375 NOK
215-4267 215-4269 215-4271
VWR®, Laboratory Bottles, Coated
Flasker Glassflasker
Made from amber borosilicate 3.3 glass, with transparent, long life plastic coating. The coating protects the bottle from scratches, splinters and leaking liquids.
  • Very good chemical resistance
  • Autoclaving or steam sterilisation at 121 or 134 °C/20 minutes
  • Frequent autoclaving reduces splintering protection
  • Long-term temperature resistance up to 140 °C/>30 minutes
  • Amber coloured, ideal for use with light-sensitive liquids
  • Delivered with mounted pouring ring and blue PP screw cap
The economical alternative for a wide range of laboratory applications.More Product Information