Over-caps, 45 mm, black phenolic, DURAN®
Leverandør: DWK Life Sciences
292492901
215-4283EA 490 NOK
215-4283
Over-caps, 45 mm, black phenolic, DURAN®
Lokk Flaskelokk
The 45 mm phenolic over-cap screws down over the open-top aluminium GL 45 cap covering and protecting the septum or stopper.
  • Made from high-quality phenol formaldehyde resin (PF)
  • High quality components
  • Recommended usable temperature is 150 °C
The phenolic cap protects the septum or stopper from damage and contamination during storage or shipping. Phenolic caps provide a wide range of chemical compatibility and temperature tolerance.