Screw cap, GL 45, PMP, DURAN® YOUTILITY®
Leverandør: DWK Life Sciences
292292843
215-4460EA 490 NOK
215-4460
Screw cap, GL 45, PMP, DURAN® YOUTILITY®
Lokk Flaskelokk
A highly transparent and ergonomic polymethylpentene cap for GL 45 DURAN® laboratory bottles.
  • High transparency (low haze) to visible light that is equivalent to DURAN® borosilicate glass
  • Compatible with all glass lab bottles and flasks with a GL 45 thread
  • Autoclavable at 121 or 134 °C, so is suitable for all GL 45 culture bottles and flasks
  • Retrace Code for traceability
  • Convenient liner-less YOUTILITY® cap design with ergonomic external grip
  • Safe with excellent chemical resistance - non toxic, BFA-free and halogen-free, food contact safe polymer (broadly