VWR® Scintillation Vials
Leverandør: VWR Collection
215-4488EA 1375 NOK
215-4488 215-4491
VWR® Scintillation Vials
Vialer
VWR® scintillation vials are produced from borosilicate 3.3 glass. They are compatible with all scintillation counters.
  • High resistance against chemicals and thermal shock
  • Low background counts and high ultraviolet transmission
  • Delivered in convenient pack sizes
VWR® scintillation vials are ideal for use in liquid scintillation counting, beta/gamma counting or for sample storage.
Bestillingsinformasjon: Screw caps have to be ordered separately and you can choose the right screw cap for your application from our selection of fitting screw caps withMore Product Information