Flasker, smal munn, HDPE/PPCO, med skrulokk, firkantet, Nalgene®
Leverandør: Thermo Fisher Scientific
2016-0030 2016-0060 2016-0125 2016-0250 2016-1000 2018-0030 2018-0060 2018-0125 2018-0250 2018-0500 2018-1000 2016-0500
215-7621EA 410 NOK
215-7621 215-7622 215-7623 215-7624 215-7626 215-7641 215-7642 215-7643 215-7644 215-7645 215-7646 215-7625
Flasker, smal munn, HDPE/PPCO, med skrulokk, firkantet, Nalgene®
Flasker Plastflasker
HDPE, gjennomskinnelig eller PPCO, gjennomskinnelig, med PP skrukork.
  • HDPE: Good chemical resistance/PPCO: Very good chemical resistance
  • HDPE: Heat resistant up to 120 °C/PPCO: Heat resistant up to 135 °C, autoclavable
  • Leakproof
Ideal for storing and shipping liquids. Graduated.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact,Product Transparency and Disclosure Low Manufacturing Impact
Miljøsertifikat NAR Certificate of Retirement, ACT Label NAR Certificate of Retirement