Disperse Yellow 3 30% (dye content), Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Warning

Synonymer: 4-(2-Hydroxy-5-methylphenylazo)acetanilide, N-[4-(2-Hydroxy-5-methylphenylazo)phenyl]acetamide

215686-50G
215686-50GEA 2380 NOK
215686-50G
Disperse Yellow 3 30% (dye content), Sigma-Aldrich®
Disperse Yellow 3

Disperse Yellow 3, Dye content 30 % (4-(2-Hydroxy-5-methylphenylazo)acetanilide; N-[4-(2-Hydroxy-5-methylphenylazo)phenyl]acetamide)

Formel: C₁₅H₁₅N₃O₂
MW: 269.3 g/mol
Smeltepunkt: 268…270 °C (lit.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002377
CAS-nummer: 2832-40-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52dafa2908209e61afb6edba64596cbcaf3c5633&vid=b7d66e078fd20c8d7bc573ba477a699ca5ae9025&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR