Macron Fine Chemicals™ CHEM-SOLV®, Laboratory Glassware Cleaner
Leverandør: MACRON AVANTOR BRAND
2157-04EA 1650 NOK
2157-04
Macron Fine Chemicals™ CHEM-SOLV®, Laboratory Glassware Cleaner
Rengjøringsløsninger Detergenter og overflatevasking
CHEM-SOLV® is a non chromic acid cleaner suitable for removing silicone greases, dried blood, and other organic residues from laboratory glass and plasticware. It is environmentally friendly and safer alternative for chromic and sulphuric acid solution.
  • Free of interfering ions
  • Ideal for cleaning labware used in sensitive analytical procedures and organic analysis
  • Cleans glassware without scrubbing