VWR®, Rack for Centrifuge Tubes, Dual
Leverandør: VWR Collection
216-0832EA 1050 NOK
216-0832
VWR®, Rack for Centrifuge Tubes, Dual
Stativer for rør Tube Racks
Polystyrene.
Securely holds 15 and 50 ml centrifuge tubes.