VWR®, Faeces/Specimen Containers, Conical, with Screw Cap
Leverandør: VWR Collection
216-0892EA 1525 NOK
216-0892 216-0893
VWR®, Faeces/Specimen Containers, Conical, with Screw Cap
Diagnostiske systemer Clinical Sample Collection/Preparation Faeces Sample Collection
PP, translucent.
  • Frosted label, graduated
  • Available with or without spoon
  • Available as sterile or packed under aseptic conditions
Ideal as faeces or general specimen containers.
Forpakning: Not individually wrapped.