Vacuum aspiration systems with trap flask

Leverandør: GRANT INSTRUMENTS
FTA-1
216-1098EA 6975 NOK
216-1098
Vacuum aspiration systems with trap flask
Pumper Vakuum remdrevet pumper
All in one system with integrated pump designed for routine aspiration of the supernatent alcohol/buffer from the walls of microtubes during DNA/RNA purification and other macromolecule reprecipitation techniques. An ideal personal tool for independent operation away from an in-line lab vacuum supply.

  • Trap flask volume 1 L
  • Fitted with hydrophobic microbiological filter
  • Small, compact and easy to use
  • Perfect for small volume aspiration
  • The hydrophobic microbiological suction filter eliminates the risk of contamination from the trap flask
  • Built-in micro-
More Product Information
Bestill nå

SPESIFIKASJONER

Modell FTA-1
Environmentally Preferable
Trykk for vakuumpumpen 500 mbar
Dimensjoner 160×210×340 mm
Gjennomstrømningshastighet 600 ml/min
Vekt 1,7 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR