Aquarium tanks
Leverandør: MARIENFELD
4204108 4204145 4204146 4204155 4204126 4204119 4204107
216-1418EA 720 NOK
216-1418 216-3830 216-3832 216-3836 216-1592 MARI4204119 MARI4204107
Aquarium tanks
Store beholdere Akvarietanker
Aquarium tanks for a large range of applications. Made from clear soda glass.
  • Strength, thick-walled
  • Ground rim
  • For applications without large demands on temperature stability