Bowls, disposable
Leverandør: Sampling Systems
8017H-05 8017H-05S
216-1554EA 4725 NOK
216-1554 SAMP8017H-05S
Bowls, disposable
Skåler Laboratoriebrett/-skåler
HDPE, white.
  • Moulded and packed in a cleanroom
  • Individually bagged
  • BSE- and TSE-free
  • Full batch traceability
Ideal for handling and mixing powders and liquids. With spout and grip.
Sertifiseringer: FDA acceptable CFR 177.1520, EU Regulations 10/2011 compliant