Single wafer shipper, for 300 mm wafer handling, H 93 series
Leverandør: ENTEGRIS
H931201011116
216-1588EA 0 NOK
216-1588
Single wafer shipper, for 300 mm wafer handling, H 93 series
Wafer håndtering Wafer forsendere
The H 93 single wafer shipper is designed to securely retain the wafer and minimise particle generation. Stackable with multiple closures, each shipper holds one 300 mm wafer facedown contacting only the wafer’s edges.
  • Secure, replaceable cushions for minimised particle generation
  • Single latch for secure closing and easy opening
  • Stackable for storage efficiency
  • Clear PC construction for visual inspection without opening
Provides secure wafer protection in a convenient, easy to use package.
Please contact Avantor for complete range of products andMore Product Information