VWR®, Straight Sample Containers, with Screw Cap
Leverandør: VWR Collection
216-1823EA 4400 NOK
216-1823 216-1808 216-1809 216-1822 216-1805 216-1806 216-1807 216-1824 216-1825 216-1826 216-1827 216-1828 216-1829 216-1830 216-1834 216-1835 216-1836 216-1837
VWR®, Straight Sample Containers, with Screw Cap
Prøvecontainere Beholdere, generelt bruk
PP, clear, with LDPE screw cap.
  • Excellent chemical and thermal resistance
  • Available sterile or packed under aseptic conditions
  • Four volumes are available: 40, 60, 125 and 180 ml
Straight walled containers for sample collection, storage and transport. Twist closure with leakproof inner lip for quick opening and closing. Screw caps assembled.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904.