VWR®, Filling and Venting Cap for VWR® Carboys
Leverandør: VWR Collection
216-1842EA 905 NOK
216-1842
VWR®, Filling and Venting Cap for VWR® Carboys
Lokk Lokk, sikkerhetsavfall
The filling and venting cap for VWR® carboys is an autoclavable closure designed for the aseptic transfer of media, biological reagents, pure water and chemicals
Each cap is equipped with integrated 6,35 mm ports.