VWR®, Jerrican, Narrow Mouth, HDPE, Rectangular
Leverandør: VWR Collection
216-1917EA 795 NOK
216-1917
VWR®, Jerrican, Narrow Mouth, HDPE, Rectangular
Flasker Jerrikanne
Disposable jerrican, made of HDPE, with integrated handle for easy carrying and pouring.
  • Good chemical resistance
  • Leakproof on standard test conditions
  • Graduated to identify the volume dispensed
  • Angled neck opening for easy pouring and transferring liquid
Jerricans are ideal for storage and transfer of sterile fluids, as well as pharmaceutical and biotechnology reagents.
Sertifiseringer: Resin meets US FDA 21 CFR, USP Class VI.