Multi-purpose trolleys
Leverandør: MULTIROIR
65007HS808E 65005HS808E3 65006HS808A3 65003HS808LE 65001HS808LE3 65002HS808LA3 GX032D GX032C
216-2231EA 4775 NOK
216-2231 216-2232 216-2234 216-2235 216-2236 216-2238 MULCGX032D MULCGX032C
Multi-purpose trolleys
Traller
These multi-use trolleys have been designed for intensive use. They are made of an aluminium frame with heavy duty polypropylene trays.
  • Grey or black
  • 2 or 3 tray versions
  • Unequalled load capacity: 80 kg per tray and 250 kg per trolley
  • 4 ball bearing wheels diam. 100 mm, 2 with brakes
The trolleys are light, easy to handle and very robust.
Bestill nå
SPESIFIKASJONER
Modell 65007HS808E 65005HS808E3 65006HS808A3 65003HS808LE 65001HS808LE3 65002HS808LA3 GX032D GX032C
Environmentally Preferable
Materiale PP PP PP PP PP PP ABS ABS
Dimensjon skål 680W×450D mm (ext.)
670W×435D mm (int.)
615W×450D mm (ext.)
607W×435D mm (int.)
615W×450D mm (ext.)
607W×435D mm (int.)
920W×520D mm (ext.)
910W×504D mm (int.)
920W×520D mm (ext.)
910W×504D mm (int.)
920W×520D mm (ext.)
910W×504D mm (int.)
645W×400D mm (ext.)
565W×325D mm (int.)
865W×490D mm (ext.)
765W×390D mm (int.)
Høyde 553 mm (height between two trays) 553 mm (height between two trays) 275 mm (height between two trays) 553 mm (height between two trays) 553 mm (height between two trays) 275 mm (height between two trays) 278 mm (height between two trays) 278 mm (height between two trays)