Jars, wide mouth, with screw cap
Leverandør: SEMADENI
1832
216-7355EA 505 NOK
216-7355
Jars, wide mouth, with screw cap
Krukker Bokser, generell bruk
Jars, wide mouth, made of HDPE.
  • Space saving
  • With white screw cap
  • Good chemical and temperature resistance
Suitable for the storage of powders, granules and viscous media.