Krukker, universal, med lokk, Nalgene®
Leverandør: Thermo Fisher Scientific
DS5300-0507 DS5300-9609 DS5300-9910
216-8262EA 1825 NOK
216-8262 216-8264 216-8265
Krukker, universal, med lokk, Nalgene®
Krukker Bokser, generell bruk
Polycarbonate, clear.
  • Drill, tap or apply clamps to jars to attach heating or cooling equipment
  • Very durable
  • Autoclavable
Multipurpose jars with cover, which can be used as water baths, aquaria/terraria or freeze-drying chambers.