Patent blue V sodium salt for mikroskopi C.I. 42051, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

21605-100G 21605-10G 21605-25G
21605-100GEA 2200 NOK
21605-100G 21605-10G 21605-25G
Patent blue V sodium salt for mikroskopi C.I. 42051, Sigma-Aldrich®
Patent blue V sodium salt

Patent blue V sodium salt has been used as a stain in pharmacology. It is mainly used as a food color.


Synonyms: Acid blue 3 sodium salt; food blue 5 sodium salt

Formel: C₂₇H₃₁N₂NaO₇S₂
MDL nummer: MFCD09608138
CAS-nummer: 20262-76-4
EINECS: 243-654-4

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR