BCECF AM ester ≥85% (by HPLC), Calbiochem®, Sigma-Aldrich®

Leverandør: Merck Millipore Calbiochem

Synonymer: BCECF AM

M-Clarity™ quality level = MQ100

216254-1
216254-1EA 3675 NOK
216254-1
BCECF AM ester ≥85% (by HPLC), Calbiochem®, Sigma-Aldrich®
BCECF AM ester

A variable mixture of cell-permeable ester derivatives of BCECF that are hydrolyzed by cytosolic esterases to yield intracellularly trapped indicator BCECF. Has been used to monitor intracellular pH changes in mammalian fibroblasts, gastric cells, lymphocytes, myocytes, and distal convoluted tubule


Forpakning: Packaged under inert gas

Formel: C₄₂H₄₀O₂₁
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00036969
CAS-nummer: 117464-70-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR