Carboxymethyl cellulose sodium salt, 800 - 3100 mPas (2%, H₂O, 25°C), white solid, high viscosity, Millipore®

Leverandør: Merck Millipore Calbiochem

217274-250
217274-250EA 1275 NOK
217274-250
Carboxymethyl cellulose sodium salt, 800 - 3100 mPas (2%, H₂O, 25°C), white solid, high viscosity, Millipore®
Carboxymethyl cellulose sodium salt

Substrate for cellulase

Formel: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00081472
CAS-nummer: 9004-32-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR