Cell scrapers
Leverandør: Biotium
22003
22003.EA 4950 NOK
22003.
Cell scrapers
Celleskrapere
PE, disposable, sterile. Mini cell scrapers for harvesting cells or cell lysates from microwell plates.
Forpakning: Packaged in 20 sterile packs of 10 scrapers per pack.