Biuret reagent solution for protein (total) in serum or plasma
Leverandør: VWR Chemicals
Warning
220947SEA 2775 NOK
220947S
Biuret reagent solution for protein (total) in serum or plasma
Biuret reagent solution
Contents per litre:
3 g Cupric sulphate CuSO4.5H2O
5.0 g Potassium iodide KI
9.0 g Potassium sodium tartrate KNaC4H4O6.4H2O
8 g Sodium hydroxide NaOH
1,4 ml Wetting agent ARW-7
Lagringstemperatur: Romtemperatur
FN: 3266
ADR: 8 II
Bestill nå
Spesifikasjon Testresultater
Suitability Passes test