VWR®, Disposable Paper Funnel
Leverandør: VWR Collection
221-0274EA 615 NOK
221-0274
VWR®, Disposable Paper Funnel
Trakter Filtreringstrakter Filtertrakter
Made of clay-coated heavy paper stock.
  • Innovative 'backrest' allows hands-free use when pouring viscous liquids
  • Fits a wide variety of openings
  • Maximum flexibility - Cut stem to any opening size or shape
  • Apply a minimum of pressure and the funnel will stay in position
  • Use with virtually any liquid or powder*
Good alternative to disposable plastic funnels. Environmentally friendly paper product will naturally biodegrade in the standard municipal waste stream. Helps to reduce cross contamination.
NB: * Not recommended for hot liquids.