Powder funnels, PP
Leverandør: Sampling Systems
8023P-100 8023P-150 8023P-250 8023P-350
SAMP8023P-100EA 53 NOK
SAMP8023P-100 221-0317 SAMP8023P-250 SAMP8023P-350
Powder funnels, PP
Trakter Pulvertrakter
Reusable powder funnel, made of PP, crevice free, with wide diameter discharge spout to enable powder to be discharged without catching.
  • Good chemical resistance
  • Good temperature resistance
  • Ideal for loading powder into drums, containers or kegs