Siphon
Leverandør: BURKLE
5605-1000
223-1513EA 1775 NOK
223-1513
Siphon
Pumper Fatpumper
Siphon pump in LDPE/PVC.
  • Good view of medium through the transparent tubing
  • With stop valve
  • Ideal for the occasional decanting of liquids
Easy to use model designed for the rapid decanting of aqueous liquids from elevated containers to those positioned lower down. Operate according to the siphon principle: Liquid needs only to be primed once. Not suitable for decanting strong acids and alkalis.