Kalsiumklorid dihydrat 97.0-103.0% Ph. Eur.

Leverandør: VWR Chemicals
Warning

22322.295EA 890 NOK
22322.295 22322.364 22322.466
Kalsiumklorid dihydrat 97.0-103.0% Ph. Eur.
Kalsiumklorid dihydrat

Appearance: White or almost white, crystalline powder, hygroscopic


NB: Not for use as Active Pharmaceutical Ingredient (API)

Formel: CaCl₂·2H₂O
Kokepunkt: 1600 °C (1013 hPa)
Smeltepunkt: 176 °C
Tetthet: 1,85 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149613
CAS-nummer: 10035-04-8
EINECS: 233-140-8
Merck Indeks: 13,01660

Bestill nå

Spesifikasjon Testresultater

Assay 97.0 - 103.0 %
Appearance Conforms (see CoA/CoS)
Identification (Cl) Passes test
Identification (Ca) Passes test
Appearance of solution Passes test
Acidity or alkalinity Passes test
SO4 (Sulphate) ≤300 ppm
Al (Aluminium) Passes test
Fe (Iron) ≤10 ppm
Magnesium and alkali metals ≤0.5 %
Heavy metals (as Pb) ≤20 ppm
Residual solvents Passes test
As (Arsenic) ≤2 ppm
Cd (Cadmium) ≤1 ppm
Co (Cobalt) ≤1 ppm
Hg (Mercury) ≤2 ppm
Ni (Nickel) ≤1 ppm
Pb (Lead) ≤5 ppm
V (Vanadium) ≤1 ppm
Conforms to Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR