Carboxymethyl cellulose sodium salt, TECHNICAL, medium viscosity

Leverandør: VWR Chemicals

Synonymer: Carboxymethyl cellulose, sodium salt, Cellulose gum, Cellulose, carboxymethyl ether, sodium salt, Sodium carboxymethyl cellulose (Viscosity 800mpa.s), Sodium carboxymethyl cellulose, CMC

22525.296EA 1925 NOK
22525.296
Carboxymethyl cellulose sodium salt, TECHNICAL, medium viscosity
Carboxymethyl cellulose sodium salt
Formel: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
Smeltepunkt: >300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00081472
CAS-nummer: 9004-32-4

Bestill nå

Spesifikasjon Testresultater

Viscosity (25°C; 2 %; water) 300 - 600 mPa.s
Water Max. 8 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR