Carborundum, TECHNICAL 0,037 mm

Leverandør: VWR Chemicals

22540.298EA 2625 NOK
22540.298
Carborundum, TECHNICAL 0,037 mm
Carborundum
Formel: SiC
Kokepunkt: < 2700 °C (1013 hPa)
Smeltepunkt: 2700 °C
Tetthet: 3,23 g/cm³ (20 °C)
CAS-nummer: 409-21-2
EINECS: 206-991-8
Merck Indeks: 12,08636

Bestill nå

Spesifikasjon Testresultater

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR