Kalsiumhydroksid ≥95%, TECHNICAL

Leverandør: VWR Chemicals
Danger

Synonymer: Slaked lime, Calcium dihydroxide

22641.460EA 3700 NOK
22641.460
Kalsiumhydroksid ≥95%, TECHNICAL
Kalsiumhydroksid
Formel: Ca(OH)₂
MW: 74.09 g/mol
Smeltepunkt: 550
Tetthet: 2.24 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010901
CAS-nummer: 1305-62-0
EINECS: 215-137-3
FN: 3262
ADR: 8 III
Merck Indeks: 13,01675

Bestill nå

Spesifikasjon Testresultater

Assay (at filling) Min. 95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR