Kloroform, Wasser_0,01 99,0-99,6 Stabilised, GPR RECTAPUR® zS

Leverandør: VWR Chemicals
Danger

Synonymer: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

Stabilisering: 0.6 % EtOH

22710.321EA 1475 NOK
22710.321
Kloroform, Wasser_0,01 99,0-99,6 Stabilised, GPR RECTAPUR® zS
Kloroform
Formel: CHCl₃
MW: 119.38 g/mol
Kokepunkt: 61.7 °C (1013 hPa)
Smeltepunkt: –63 °C
Tetthet: 1.4832 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000826
CAS-nummer: 67-66-3
EINECS: 200-663-8
FN: 1888
ADR: 6.1 III
REACH: 01-2119486657-20
Merck Indeks: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay 99.0 - 99.6 %
Density (20/4) 1.473 - 1.489
E/E 0.1 - 1.0 %
Evaporation residue Max. 50 ppm
Water Max. 100 ppm

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR