Kolesterol, Millipore®

Leverandør: Merck Millipore Calbiochem

228111-5
228111-5EA 1575 NOK
228111-5
Kolesterol, Millipore®
Kolesterol
Formel: C₂₇H₄₆O
Kokepunkt: 360 °C (1013 hPa)
Smeltepunkt: 147…150 °C
Tetthet: 1,07 g/cm³ (20 °C)
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00003646
CAS-nummer: 57-88-5
EINECS: 200-353-2
Merck Indeks: 13,02221

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR