Slangeklemme
Leverandør: Bel-Art Products, a Part of SP
F182120000
229-0007EA 775 NOK
229-0007
Slangeklemme
Klemmer Slangeklemmer
Molded of Acetal plastic. This tubing clamp offers on/off and fine control so accurate that it replaces stopcocks in many applications.
  • Can be installed without disconnecting tubing or interrupting operation
  • Accepts flexible tubing up to 14 mm Ext.Ø
  • Resistant to acids, alkalis and organic solvents
  • Autoclavable