Tubing clips with screw thread
Leverandør: KOHLER
1267702032
229-7318EA 260 NOK
229-7318
Tubing clips with screw thread
Klemmer Slangeklemmer
Stainless steel tightening strap, rust-proof with galvanised thread screw.
  • Good chemical resistance, excellent temperature resistance, acid-resistant
  • Secure and tight fit
  • Easy to assemble and disassemble using a slot screwdriver