Solvent Blue 59 98% (dye content), Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Warning

Synonymer: Sudan Blue, Atlasol Blue 2N, 1,4-Bis(ethylamino)-9,10-anthraquinone

229121-25G
229121-25GEA 1210 NOK
229121-25G
Solvent Blue 59 98% (dye content), Sigma-Aldrich®
Solvent Blue 59

Solvent Blue 59, Dye content 98 % (1,4-Bis(ethylamino)-9,10-anthraquinone; Atlasol Blue 2N; Sudan Blue)

Formel: C₁₈H₁₈N₂O₂
MW: 294.35 g/mol
Smeltepunkt: 215…217 °C (lit.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00071821
CAS-nummer: 6994-46-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9326389d5952d2afc9e3ded699b9283319dbc543&vid=77af2c57d72e6899d07bc69021c26a9ea0d784c9&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR