N,N-Dimetylacetamid ≥99.5% for HPLC

Leverandør: Thermo Scientific
Danger

Synonymer: Acetic acid dimethylamide, Dimethyl Acetamide, DMAC

22916.K2
22916.K2EA 1700 NOK
22916.K2
N,N-Dimetylacetamid ≥99.5% for HPLC
N,N-Dimetylacetamid

Purity: 99.5+% CAS No.: 127-19-5 Molecular Formula: C4H9NO Formula Weight: 87.12 Flash Point: 70°(158°F) Physical Form: Liquid Density: 0.940 Refractive Index: 1.4385 Appearance: Colourless Odour: Ammonia-like Melting Point: -20° Boiling Point: 165-166° Beilstein Reference: 1737614 Merck Reference: 14,3227 MDL No.: MFCD00008686

Formel: CH₃CON(CH₃)₂
MW: 87,12 g/mol
Kokepunkt: 165…166 °C
Smeltepunkt: –20 °C
Tetthet: 0,940
Flash punkt: 70 °C (158 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008686
CAS-nummer: 127-19-5
EINECS: 204-826-4
FN: 2810
ADR: 6.1,III
Merck Indeks: 14,3227
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=4e98bd4c34a9326a56ee3e11797b2e6e1b323c8f&prid=web&clid=SAPDAM

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR