Carminic acid 90 HPLC_ASSAY_METHOD, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY


MQ100

229253-1G 229253-5G
229253-1GEA 950 NOK
229253-1G 229253-5G
Carminic acid 90 HPLC_ASSAY_METHOD, Sigma-Aldrich®
Carminic acid

Carmine is a cochineal product usually furnished as an aluminum and calcium salt of carminic acid. Carminic acid is used for human-medical cell diagnosis and serves the purpose of the histological investigation of sample material of human origin. It is suitable for staining nuclei in histological sections. It is utilised in various staining methods including the procedure for detection of glycogen and mucous substances (Best′s carmine), for nuclear staining, or vital staining.


Carminic acid (CI 75470) is used in Orth′s lithium carmine technique for nuclear staining, Best′s stain for glycogen, and Southgates′s mucicarmine method.

Formel: C₂₂H₂₀O₁₃
Smeltepunkt: 136 °C
Lagringstemperatur: Room Temperature
MDL nummer: MFCD00167028
CAS-nummer: 1260-17-9
EINECS: 215-023-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7983dbd26ace2478956601cdd32bbbfc01fb2615&vid=ff95adec14aa8f66d877b9a624d35982f943e33f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance (Color) Red to Very Dark Red and Purple and Red-Purple and Purple-Red
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Wavelength c = 0.03g/L; Ethanol 494 - 500 nm
Extinction Coefficient ≥13000
Wavelength 222 - 228 nm
Purity (HPLC) ≥90.0 %
Solubility (Turbidity) c = 1%; 1N Acetic Acid Clear to Hazy
Solubility (Color) Red to Very Dark Red


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR