Benedict's reagent for qualitative detection of sugar in urine

Leverandør: VWR Chemicals

230045REA 1775 NOK
230045R
Benedict's reagent for qualitative detection of sugar in urine
Benedict's Reagent

Benedict’s reagent for qualitative detection of sugar in urine.

MDL nummer: MFCD00146068
CAS-nummer: 63126-89-6

Bestill nå

Spesifikasjon Testresultater

S.G. @ 20°C 1.192 - 1.195
Dextrose Test Positive Reaction
Thiocyanate Test Negative Reaction

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR