Flow cytometry fixation/permeabilisation kit
Leverandør: Biotium
23006
23006.EA 615 NOK
23006.
Flow cytometry fixation/permeabilisation kit
Assay Cellulær analyse
Flow Cytometry Fixation/Permeabilisation Kit contains optimally formulated buffers for fixation and permeabilisation of suspension cells for immunofluorescence staining of intracellular antigens for analysis by flow cytometry. The bufffers are also available separately.
Leveringsinformasjon: Kit contains Fixation Buffer, 5 ml and Permeabilisation Buffer, 5 ml.