Sample collection system (scoop and bag), SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
H369100000
231-0011EA 3250 NOK
231-0011
Sample collection system (scoop and bag), SP Bel-Art
Øser
PS, white. This sample collection system includes a sterile 60 ml (2 oz.) sampling scoop and a sterile 540 ml (18,3 oz.) volume PE twirl-type bag with large label area to hold the sample.
  • One-piece scoop made from white, high-impact, FDA grade PS
  • Long handle (127 mm) keeps the hand away from the sample
  • Flat bottom allows scoop to remain upright for use as a weighing boat or to be placed on a flat surface
  • 50 individually sealed and sterilised wrapped sets per box ready for one time use and disposal sterile
  • Lot stamped for accurate lot tracing; shipped with