Spoon spatulas, for analysis
Leverandør: BOCHEM
3219
231-0045EA 120 NOK
231-0045
Spoon spatulas, for analysis
Spatler
Stainless steel ¹⁸/₁₀, anti-magnetic. Multipurpose spatula for a wide range of uses.
  • With one tapered end
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Anti-magnetic, electrolytically polished