Digeltenger
Leverandør: NICKEL ELECTRO
3021
231-0353EA 0 NOK
231-0353
Digeltenger
Tenger
Pure nickel (99,6%). With grooved tips. For easy, safe handling of crucibles made from a wide variety of materials.
  • Excellent corrosion resistance
  • Good thermal and electrical conductivity
  • Excellent thermal resistance