Digeltenger
Leverandør: NICKEL ELECTRO
3041
231-0354EA 125 NOK
231-0354
Digeltenger
Tenger
Steel, non alloy. For easy, safe handling of crucibles made from a wide variety of materials.
  • With grooved tips
  • Good thermal resistance
  • Robust and easy action