Øser, PharmaScoop
Leverandør: BURKLE
5324-0016 5324-0015 5324-0012 5324-0013 5324-0014 5324-0017 5324-0018 5324-0010 5324-0011
300-0152EA 5825 NOK
300-0152 300-0151 300-0148 300-0149 300-0150 300-0153 300-0154 BURK5324-0010 231-0496
Øser, PharmaScoop
Øser
Stainless steel AISI 316 (1,4401). This sample scoop has been developed in order to meet the high requirements of the pharmaceutical industry, such as those contained in the FDA regulations.
  • GMP conform
  • Highly polished
  • Without edges, crevices or undercuts in which sample residues could collect and contaminate subsequent samples
The scoops meet all the requirements for GMP compliance in production and sampling.