VWR®, Scoop
Leverandør: VWR Collection
231-0501EA 495 NOK
231-0501
VWR®, Scoop
Øser
PTFE, white.
  • Totally chemically inert
  • Non contaminating
  • Suitable for use with frozen substances and other solids
Scoop length: 80 mm
Total length: 140 mm