Microspatulas, LaboPlast® / SteriPlast®
Leverandør: BURKLE
5378-0032 5378-1032
231-0606EA 675 NOK
231-0606 231-0607
Microspatulas, LaboPlast® / SteriPlast®
Spatler
PS. Double-scoop spatulas with rounded ends, content 1,0 and 0,25 ml. Can be used to sample, transfer or process small amounts of chemicals, powders, granulates, pastes, creams or liquids.
  • Sturdy and convenient handle facilitates precise work
  • Production, assembly and packaging according to cleanroom Class 7
  • Complies with EU food requirements and FDA regulations
  • Optional individual packaging sterilised by gamma rays
Forpakning: Available either in a large packs with 100 pieces or 100 pieces packed individually and sterilised by gamma rays.